Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
25 g | Please contact Customer Service for Availability | 2 430,00 CZK |
100 g | Please contact Customer Service for Availability | 8 150,00 CZK |
About This Item
2 430,00 CZK
form
(Powder to crystal)
Quality Segment
technique(s)
cell culture | mammalian: suitable
mp
67-71 °C
solubility
soluble (50mg/ml)
shipped in
wet ice
storage temp.
2-8°C
SMILES string
Cl[H].NCCS
InChI
1S/C2H7NS.ClH/c3-1-2-4;/h4H,1-3H2;1H
InChI key
OGMADIBCHLQMIP-UHFFFAOYSA-N
General description
Application
Features and Benefits
1 of 1
This Item | |||
|---|---|---|---|
| description suitable for cell culture | description ≥97.0% (RT) | description EMPROVE® ESSENTIAL | description ≥98% (titration) |
| solubility soluble (50mg/ml) | solubility - | solubility - | solubility water: 50 mg/mL, clear, colorless to very faintly yellow |
| Quality Level 200 | Quality Level 200 | Quality Level 500 | Quality Level 300 |
| form (Powder to crystal) | form crystals | form solid | form powder or crystals |
| mp 67-71 °C | mp 64-70 °C, 67-71 °C | mp 67-71 °C | mp 67-71 °C |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| shipped in wet ice | shipped in - | shipped in - | shipped in - |
Still not finding the right product?
Explore all of our products under Cysteamine hydrochloride
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

