Sign In to View Organizational & Contract Pricing.
About This Item
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC
Citations:
5
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
immunocytochemistry: 1 μg/mL using HEK293 cells overexpressing human CFTR
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CFTR(1080)
rat ... Cftr(24255)
General description
CFTR (Cystic fibrosis transmembrane conductance regulator, ATP-binding cassette sub-family C, member 7) is a membrane-associated, N-linked glycoprotein.
Immunogen
synthetic peptide corresponding to amino acids 103-117 of human CFTR protein. The sequence is 100% conserved in human, rabbit, and monkey. There is a one amino acid substitution in rat, bovine, and sheep.
Application
Anti-CFTR antibody produced in rabbit is suitable for immunocytochemistry at a concentration of 1μg/mL using HEK293 cells overexpressing human CFTR.
Biochem/physiol Actions
Cystic fibrosis transmembrane conductance regulator (CFTR) gene is associated with congenital bilateral absence of the vas deferens (CBAVD) and causes the genital form of cystic fibrosis (CF). The CFTR gene may also be involved in the etiology of male infertility. It may be useful for assisting reproduction technology. There is nearly 2000 cystic fibrosis transmembrane regulator (CFTR) mutations that are associated with cystic fibrosis (CF).
Physical form
Solution in phosphate buffered saline containing 1.0 mg/mL bovine serum albumin and 0.05 % sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Explore all of our products under Anti-CFTR antibody produced in rabbit
— or —
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.