Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
manufacturer/tradename
IROA Technologies
packaging
pkg of 1 plate
suitability
suitable for LC-MS, suitable for mass spectrometry
application(s)
lipidomics
metabolomics
(microbiome)
relevant disease(s)
cardiovascular diseases; neurological disorders; metabolic disorders; endocrinological disorders
shipped in
dry ice
storage temp.
−20°C
General description
Application
Features and Benefits
2. Aids in optimizing the mass spectrometry analytical protocols, to qualify and quantify mass spectrometry sensitivity.
3. Consists of high purity compounds that are supplied in an economical, 96-well plate.
4. Pre-weighed, and supplied in dry and ready to use format.
5. Available plate map of compounds and descriptors provided in a downloadable spreadsheets:
Preparation Note
Other Notes
• Branched-chain acylcarnitines
• Hydroxylated carnitines
• Unsaturated acylcarnitines
• Short-chain acylcarnitine
• Medium-chain acylcarnitines
• Long-chain acylcarnitines
CARMLS is distributed with MLSDiscovery™, a software tool developed by IROA Technologies to support the extraction, manipulation, and storage of spectral data generated during library-based metabolomics workflows. The software enables rapid construction of mass spectral libraries using the user′s own analytical conditions, capturing retention time, mass, and relative intensity information for compounds, fragments, and adducts.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description Supplied by IROA Technologies | description Supplied by IROA Technologies | description Supplied by IROA Technologies | description Supplied by IROA Technologies |
| application(s) lipidomics | application(s) - | application(s) - | application(s) - |
| manufacturer/tradename IROA Technologies | manufacturer/tradename IROA Technologies | manufacturer/tradename IROA Technologies | manufacturer/tradename IROA Technologies |
| packaging pkg of 1 plate | packaging pkg of 1 plates | packaging pkg of 2 plates | packaging pkg of 1 plates |
| relevant disease(s) cardiovascular diseases; neurological disorders; metabolic disorders; endocrinological disorders | relevant disease(s) - | relevant disease(s) neurological disorders; metabolic disorders | relevant disease(s) - |
| suitability suitable for mass spectrometry, suitable for LC-MS | suitability - | suitability - | suitability - |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



