Skip to Content
Merck

Skip To

CLVIII-3A6-E3 Mouse x Mouse Hybridoma, ECACC 08041720

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41106514
Biological source:
Mouse x Mouse Hybridoma
Isotype:
IgG2b
Technique(s):
cell culture | mammalian: suitable
Myeloma:
NS-1 (P3-X63/Ag8-NS1)
Reactivity:
Anti-L. panamensis-Amastigote & Promastigote
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

CLVIII-3A6-E3, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

Mouse x Mouse Hybridoma

manufacturer/tradename

ECACC

packaging

tube of 5 μg 08041720-DNA-5UG, pkg of vial of cells 08041720-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

NS-1 (P3-X63/Ag8-NS1)

Reactivity

Anti-L. panamensis-Amastigote & Promastigote

technique(s)

cell culture | mammalian: suitable

isotype

IgG2b

shipped in

dry ice

storage temp.

−196°C

Immunogen

Membrane from Leishmania (V) panamensis strain WR442

Application

Detection of Leishmania (V) panamensis

Biochem/physiol Actions

CLVIII-3A6-E3 was developed by immunizing mice with membranes from L. panamensis strain WR442 and fusing the mouse spleen cells with NS-1(P3-X63/Ag8-NS1)
Mouse NS1 x BALB/c mouse

Preparation Note

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Seed cultures between 1-3x100,000 cells/ml; Saturation density 1,000,000 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CB_08041712CB_08041724CB_08041715
vibrant-m

CLVIII-3A6-E3

vibrant-m

LXVIII-4D8-E3

vibrant-m

XCIII-3D8-D8

vibrant-m

XXIX-1G12-G7

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

packaging

tube of 5 μg 08041720-DNA-5UG, pkg of vial of cells 08041720-1VL

packaging

pkg of vial of cells 08041712-1VL, tube of 5 μg 08041712-DNA-5UG

packaging

tube of 5 μg 08041724-DNA-5UG, pkg of vial of cells 08041724-1VL

packaging

tube of 5 μg 08041715-DNA-5UG, pkg of vial of cells 08041715-1VL

isotype

IgG2b

isotype

IgG1

isotype

IgG1

isotype

IgG1

ImmunoDon

BALB/c mouse spleen

ImmunoDon

Balb/c mouse spleen

ImmunoDon

Balb/c mouse spleen

ImmunoDon

BALB/c mouse spleen

Myeloma

NS-1 (P3-X63/Ag8-NS1)

Myeloma

NS-1 (P3-X63/Ag8-NS1)

Myeloma

NS-1 (P3-X63/Ag8-NS1)

Myeloma

NS-1 (P3-X63/Ag8-NS1)

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters