Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41106514
eCl@ss:
32160703
Biological source:
mouse nerve
Relevant disease(s):
cancer
Growth mode:
Adherent
Karyotype:
Not specified
Morphology:
Neuronal
Manufacturer/tradename:
ECACC
Product Name
N115-BU-9, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
biological source
mouse nerve
manufacturer/tradename
ECACC
packaging
tube of 5 μg 08062558-DNA-5UG, pkg of vial of cells 08062558-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Neuronal
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Application
Used in the generation of cell hybrids.
Biochem/physiol Actions
A bromodeoxyuridine-resistant mutant of N1E-115 (Sigma Catalogue number 08062511). The cell line N1E-115 is an adrenergic cell line derived from mouse neuroblastoma C1300 (Mouse strain A/Jax).
Mouse Neuroblastoma
Preparation Note
DMEM + 2 mM glutamine + 10% Foetal Bovine Serum (FBS) + 1 mM 8-bromodeoxyuridine. The cells can be grown in the absence of bromodeoxyuridine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.