Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Product Name
COR-L64, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
biological source
human bone
manufacturer/tradename
ECACC
packaging
tube of 5 μg 96020749-DNA-5UG, pkg of vial of cells 96020749-1VL
growth mode
Suspension
karyotype
Not specified
morphology
Floating aggregates of packed cells. Irregular shaped aggregates
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Biochem/physiol Actions
Preparation Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. | description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. | description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 00060606, human tongue, squamous cell carcinoma. Epithelial-like morphology. | description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 91091803, human lung, Epithelial |
| packaging tube of 5 μg 96020749-DNA-5UG, pkg of vial of cells 96020749-1VL | packaging pkg of vial of cells 91011802-1VL, tube of 5 μg 91011802-DNA-5UG | packaging tube of 5 μg 00060606-DNA-5UG, pkg of vial of cells 00060606-1VL | packaging tube of 5 μg 91091803-DNA-5UG, pkg of vial of cells 91091803-1VL |
| karyotype Not specified | karyotype Not specified | karyotype Not specified | karyotype Not specified |
| growth mode Suspension | growth mode - | growth mode - | growth mode Adherent |
| morphology Floating aggregates of packed cells. Irregular shaped aggregates | morphology - | morphology - | morphology Epithelial |
| products Not specified | products - | products - | products Not specified |
| storage temp. −196°C | storage temp. - | storage temp. - | storage temp. −196°C |
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
