Skip to Content
Merck

D1636

Diethyl (ethoxymethylene)malonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C2H5OCH=C(COOC2H5)2
CAS Number:
Molecular Weight:
216.23
UNSPSC Code:
12352200
EC Number:
201-725-7
Beilstein/REAXYS Number:
880058
MDL number:
eCl@ss:
32150211
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


bp

279-283 °C (lit.)

density

1.080 g/mL at 20 °C (lit.)

SMILES string

CCO\C=C(\C(=O)OCC)C(=O)OCC



Still not finding the right product?

Explore all of our products under Diethyl (ethoxymethylene)malonate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library