Skip to Content
Merck

D2026

Hexamethylene diisocyanate

Synonym(s):

1,6-Diisocyanatohexane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
OCN(CH2)6NCO
CAS Number:
Molecular Weight:
168.19
EC Number:
212-485-8
UNSPSC Code:
12352103
MDL number:
Beilstein/REAXYS Number:
956709
eCl@ss:
32150319
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


bp

82-85 °C/0.1 mmHg

density

1.047 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C=NCCCCCCN=C=O

InChI

1S/C8H12N2O2/c11-7-9-5-3-1-2-4-6-10-8-12/h1-6H2

InChI key

RRAMGCGOFNQTLD-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Hexamethylene diisocyanate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library