Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CNCH2CH2CH2CH2CH2CH2NC
CAS Number:
Molecular Weight:
136.19
EC Number:
213-202-0
UNSPSC Code:
12352100
MDL number:
Beilstein/REAXYS Number:
3662465
eCl@ss:
32150313
refractive index
n
bp
101-103 °C/0.5 mmHg (lit.)
mp
21-23 °C (lit.)
density
0.899 g/mL at 25 °C (lit.)
storage temp.
−20°C
SMILES string
[C-]#[N+]CCCCCC[N+]#[C-]
InChI
1S/C8H12N2/c1-9-7-5-3-4-6-8-10-2/h3-8H2
InChI key
WJCPMQQLTJQIJK-UHFFFAOYSA-N
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Eye Irrit. 2
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point (°F)
219.2 °F - closed cup
Flash Point (°C)
104 °C - closed cup
PPE (personal protective equipment)
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
