Skip to Content
Merck

Skip To

D3572

Dipeptidyl Peptidase IV Inhibitor IV

Synonym(s):

(S)-1-(4-Methyl-1-(2-pyrimidinyl)-4-piperidylamino)acetyl-2-pyrrolidinecarbonitrile, K579

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C17H24N6O
Molecular Weight:
328.41
NACRES:
NA.77
UNSPSC Code:
12352202
Assay:
≥97% (HPLC)


assay

≥97% (HPLC)

Quality Segment

form

solid

storage temp.

2-8°C

General description

Dipeptidyl Peptidase IV Inhibitor IV, also known as K579 ((S)-1-[4-methyl-1-(2-pyrimidinyl)-4-piperidylamino]acetyl-2-pyrrolidinecarbonitrile), is orally available. It has a formula weight of 339.39 g/mol.

Biochem/physiol Actions

Dipeptidyl peptidase IV (DPP IV) is a key enzyme involved in the regulation of incretin hormone levels in the gastrointestinal tract. Incretin hormones, such as glucagon like peptide (GLP-1) and glucose-dependent insulinotropic hormone (GIP) / gastric inhibitory peptide, cause the glucose stimulated release of insulin. This allows the endocrine system to preemptively stimulate release insulin in response to ingestion of high sugar foods. Type 2 diabetics demonstrate a greatly reduced incretin induced insulin release response.
This response in hyperglycemic conditions is counterbalanced by the rapid rate of degradation of these incretin hormones by dipeptidyl peptidase IV in normal conditions. Inhibition of dipeptidyl peptidase IV results in longer lifetime of the incretin hormones in the GI tract and thus prolonged stimulation of insulin production. Specific dipeptidyl peptidase inhibitors are actively being explored as drug targets for treating type 2 diabetes and several candidates are actively in clinical trial.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters