Skip to Content
Merck

D3818

Dextran Ladder

Glycan standard for HPLC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352201
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bacterial (Leuconostoc mesenteroides)

form

solid

shipped in

wet ice

storage temp.

−20°C

Application

The dextran ladder is used as an external standard for the analysis of glycans by normal phase HPLC after fluorescent labeling. This standard gives a characteristic ladder profile from monomeric glucose to approximately a 20-mers of glucose oligosaccharide, dependent upon the chromatographic conditions employed. The elution position of each peak in this ladder is expressed as a glucose unit (gu) and is used to assign gu values to peaks in the released glycan pool.Dextran ladder upon derivitization can be used as a new tool to assess cell layer tightness.

Preparation Note

The dextran ladder is prepared by partial acid hydrolysis of dextran from Leuconostoc mesenteroides with an average molecular weight of 100-200 kDa. The purity and structural integrity of the ladder is assessed by fluorescently labeling an aliquot and subsequent analysis by normal phase HPLC.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library