Skip to Content
Merck

D5266

Sodium 1-dodecanesulfonate

Synonym(s):

1-Dodecanesulfonic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)11SO3Na
CAS Number:
Molecular Weight:
272.38
EC Number:
219-200-6
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
3919536
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

anionic

mp

>300 °C (lit.)

SMILES string

[Na+]

InChI

1S/C12H26O3S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DAJSVUQLFFJUSX-UHFFFAOYSA-M



Still not finding the right product?

Explore all of our products under Sodium 1-dodecanesulfonate


Storage Class

11 - Combustible Solids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library