Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 units | Please contact Customer Service for Availability | 2 280,00 CZK |
1500 units | Please contact Customer Service for Availability | 17 000,00 CZK |
About This Item
2 280,00 CZK
Quality Segment
biological source
mouse ascites (recombinant)
recombinant
expressed in E. coli
form
liquid
feature
dNTPs included: no, hotstart, Standard PCR
concentration
5 unit/μL
technique(s)
PCR: suitable
input
purified DNA
density
1 g/cm3
shipped in
wet ice
storage temp.
2-8°C
General description
Glycerol-free JumpStart™ Taq DNA Polymerase is suitable for lyophilization and allows the user to lyophilize PCR master mixes, which can be stored at ambient temperature and used up to one year later without reagent degradation.
Application
Features and Benefits
- Designed to minimize non-specific amplification
- Increased specificity and target yield
- Reduced primer-dimers
- Reduced set-up time as compared to manual or wax Hot Start methods
- Allows room-temperature reaction set-up
- Suitable for multiplex PCR
- Lyophilization-compatible hot-start enzyme that is glycerol-free
Packaging
Other Notes
Legal Information
Antibody licensed for in vitro research use under U.S. Patent No. 5,338,671 and 5,587,287, and corresponding patents in other countries.
1 of 1
This Item | |||
|---|---|---|---|
| description Lyophilization-compatible | description without MgCl2 | description with MgCl2 | description DNA-free, Nuclease-free Hot-start Taq Polymerase, designed for further manufacture |
| biological source mouse ascites (recombinant) | biological source - | biological source - | biological source - |
| technique(s) PCR: suitable | technique(s) PCR: suitable, whole genome amplification: suitable | technique(s) whole genome amplification: suitable, PCR: suitable | technique(s) PCR: suitable, cDNA synthesis: suitable, DNA sequencing: suitable |
| form liquid | form liquid | form liquid | form liquid |
| recombinant expressed in E. coli | recombinant - | recombinant - | recombinant expressed in E. coli expressed in |
| concentration 5 unit/μL | concentration 2.5 units/μL | concentration 2.5 units/μL | concentration 2.5 units/μL |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in dry ice |
Storage Class
12 - Non Combustible Liquids
WGK
WGK 2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



