Skip to Content
Merck

Skip To

E1272

Lyo ECM Gel

lyophilized powder, BioReagent, suitable for cell culture

Synonym(s):

Basement membrane extract (BME), EHS matrix, Extracelular matrix, ECM

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.41
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

form

lyophilized powder

concentration

8-12 mg/mL (after reconstitution)

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

General description

ECM gel is a solubilized basement membrane matrix secreted and extracted from EHS mouse sarcoma cells that works as a complex network of proteins and other molecules that surround, support, and give structure to mammalian tissues. Lyo ECM Gel is a novel lyophilized version of the normal ECM gel product providing adjustable research concentrations, enhancing product stability and ease-of-use for many biological applications. The extracellular matrix helps cells attach to, and communicate with, nearby cells. It plays an important role in cell functioning, including growth and movement. Commonly used for: Culturing of invasive cells, induce stem cells proliferation and differentiation, formation of organoids, suitable for the development of cancer cells and xenografts.

Prevention of cancer metastasis must be a major goal of cancer therapy since metastasis is the most insidious and life-threatening aspect of cancer . 3D tumor organoid systems have been developed for cancer cell types. These complex, self-organizing structures are cultured forms of tumor cells embedded and growth supported by ECM GEL.

ECM gel contains laminin as a major component, collagen type IV, heparan sulfate proteoglycan, entactin, and other minor components. ECM gel undergoes thermally activated polymerization when brought to 20-40°C to form a reconstituted basement membrane. The process of gelation is reversible. Addition of collagen type IV to ECM gel increases polymerization, whereas, addition of collagen type I, fibronectin, or heparin, does not. PC12 cells show neurite formation within 2 days when grown on a thin layer of ECM Gel.

Every mouse colony used for the production of this product is routinely screened for several pathogens. Tested and found negative for LDEV

Preparation Note

Lyo ECM Gel is recomended to be reconstituted with 10 ml of water (#W3500) for a concentration as described in COA specifications (8-12 mg/ml). Lyo ECM Gel can be storage at room temperature for up to 4 months. The reconstituted liquid Lyo ECM Gel can be keep upto 7 days at 4oC. During work is recomended to keep the vial in ice or in a temperature under 10 oC

Disclaimer

This product is for R&D use only, not for drug, household, or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CC131E0282CC176
description

lyophilized powder, BioReagent, suitable for cell culture

description

Stem cell qualified basement membrane extract (BME) for human ES and iPS cell culture.

description

mixture, liquid

description

-

form

lyophilized powder

form

gel form

form

liquid

form

-

storage temp.

2-8°C

storage temp.

-

storage temp.

−70°C

storage temp.

2-8°C

Quality Level

200

Quality Level

-

Quality Level

200

Quality Level

-

product line

BioReagent

product line

-

product line

-

product line

-

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | stem cell: suitable

technique(s)

-

technique(s)

cell culture | mammalian: suitable

concentration

8-12 mg/mL (after reconstitution)

concentration

-

concentration

-

concentration

-


Storage Class

13 - Non Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library