Skip to Content
Merck

E2406

Endoglycosidase H from Streptomyces griseus

lyophilized powder

Synonym(s):

β-N-Acetylglucosaminidase H

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
MDL number:
EC Number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized powder

specific activity

≥10 units/mg protein



mol wt

29 kDa

shipped in

dry ice

storage temp.

−20°C

Physical form

Lyophilized from a solution containing 10 mM Tris HCl, pH 7.2. This product does not contain a protein stabilizer and must be reconstituted with a solution containing BSA or another stabilizer.

Other Notes

One unit will hydrolyze 1.0 μmole of N-acetyl-(14C)Asn(GlcNAc)2(Man)5 per min at pH 5.0 at 37 °C.


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library