Skip to Content
Merck

Skip To

E5404

Anti-Ephrin-B1 antibody produced in goat

affinity isolated antibody, lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Please contact Customer Service for Availability
14 700,00 CZK

About This Item

UNSPSC Code:
51111800
NACRES:
NA.41
MDL number:
Technique(s):
western blot: 0.1-0.2 μg/mL, immunohistochemistry: 2-15 μg/mL using cells and tissues, indirect ELISA: 0.5-1.0 μg/mL
Conjugate:
unconjugated
Application:
ELISA (i), IHC, WB
Citations:
3

14 700,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

western blot: 0.1-0.2 μg/mL, immunohistochemistry: 2-15 μg/mL using cells and tissues, indirect ELISA: 0.5-1.0 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Efnb1(13641)

General description

By ELISA, the antibody shows less than 1% cross-reactivity with rhEphrin-A3, rhEphrin-A4, rhEphrin-A5, and rhEphrin-B3. Also, there is less than 1% cross-reactivity with rmEphrin-A1, rmEphrin B2, rmEphrin-A2, rmEphrin-A4, and rzfEphrin-B2.

Immunogen

purified recombinant mouse ephrin-B1 extracelluar domain expressed in mouse NSO cells.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

Ephrin B4 binds to EPH receptor B, a protein-tyrosine kinase family that belongs to the ephrin receptor subfamily. EphB receptors and EphrinB ligands have a regulatory effect on endothelial cell function, angiogenesis, migration, cell growth and survival.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline and 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I2136F8264P2860
description

affinity isolated antibody, lyophilized powder

description

affinity isolated antibody, buffered aqueous solution

description

affinity isolated antibody, buffered aqueous solution

description

clone B92, purified from hybridoma cell culture

biological source

goat

biological source

goat

biological source

goat

biological source

mouse

Quality Level

100

Quality Level

300

Quality Level

200

Quality Level

200

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

conjugate

unconjugated

conjugate

unconjugated

conjugate

FITC conjugate

conjugate

unconjugated

UniProt accession no.

P52795

UniProt accession no.

-

UniProt accession no.

-

UniProt accession no.

-

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

B92, monoclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters