Skip to Content
Merck

Skip To

E5768

Monoclonal Anti-Eotaxin-2 antibody produced in mouse

clone 61016.11, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-MPIF-2

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
51111800
NACRES:
NA.41
MDL number:
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
61016.11, monoclonal
Application:
ELISA (c), Neutral, WB
Citations:
8


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

61016.11, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

capture ELISA: suitable, neutralization: suitable, western blot: 1-2 μg/mL

isotype

IgG1

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCL24(6369)

General description

Accumulation of eosinophils is the hallmark of several medical conditions such as infections and systemic diseases. A number of chemokines stimulate the proliferation, migration and accumulation of eosinophils at the site of inflammation. Eotaxin-2 is chemoattractant that selectively acts on eosinophils and has minimal effect on other leukocytes. It is secreted in response to TNF-α, IL-5 or IL-3. The effect of Eotaxin is mediated by specific eotaxin receptor that induces activation of Gi proteins, MAPK signaling and involves changes in intracellular calcium levels and cytoskeletal reorganization. Eotaxin receptor is present on eosinophils, basophils and T-helper cells. Eotaxin coordinates the accumulation of these cells in response to allergic inflammation and homing of eosinophils to gastrointestinal tract and mucosal tissues
Monoclonal Anti-Eotaxin-2 (MPIF-2) recognizes recombinant human eotaxin-2, less than 0.03 % cross-reactivity with recombinant human eotaxin, MIP-1α, and MCP-3.

Immunogen

recombinant human eotaxin-2.

Application

Anti-eotaxin-2 antibody may be used for neutralization assays at a working concentration of 1-5 μg/ml in the presence of 100 ng/ml of human recombinant eotaxin-2. The antibody is suitable for immunoblotting at 1-2 μg/ml concentration. For capture ELISA, a final concentration of 2 μg/ml may be used.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline

Preparation Note

Purified using protein G

Analysis Note

The antibody neutralizes the biological activity of recombinant human eotaxin-2. It exhibits less than 0.03% cross-reactivity with recombinant human eotaxin, MIP-1α, and MCP-3.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters