Skip to Content
Merck

Skip To

E7282

EPHB4 (561-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

HTK, MYK1, Mdk2, TYRO11

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 μg
Please contact Customer Service for Availability
9 730,00 CZK

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
MDL number:
Specific activity:
349-472 nmol/min·mg
Assay:
≥70% (SDS-PAGE)
Recombinant:
expressed in baculovirus infected Sf9 cells

9 730,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

349-472 nmol/min·mg

mol wt

~50 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EPHB4(2050)

General description

EPHB4 (Ephb1 Eph receptor B4) belongs to EPHB receptor family, which is the biggest family of transmembrane tyrosine kinase receptors. Mammals contain five EPHB receptors, namely, EPHB1-EPHB4 and EPHB6. Their ligands are transmembrane in nature and are called ephrinB1, -B2 and -B3.

Biochem/physiol Actions

EPHB4 (ephrin receptor B4) is a member of the Eph family of receptor tyrosine kinases which play a role in regulating cell adhesion and cell movement during embryonic development. Furthermore, because tissue disorganization and abnormal cell adhesion, movement, and survival characterize the more advanced stages of cancer, the inappropriate functioning of Eph receptor could play a causal role in malignancy. Furthermore, EPHB4 has been shown to be the major essential interaction partner of Ephrin B2 in angiogenesis.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SRP0347E1158SAB5300476
description

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

description

recombinant, expressed in baculovirus infected Sf9 cells

description

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

description

clone 5B8F7, ascites fluid

specific activity

349-472 nmol/min·mg

specific activity

-

specific activity

22-30 nmol/min·mg

specific activity

-

Gene Information

human ... EPHB4(2050)

Gene Information

human ... EPHB4(2050)

Gene Information

human ... EPHB3(2049)

Gene Information

human ... EphB4(2050)

assay

≥70% (SDS-PAGE)

assay

-

assay

≥70% (SDS-PAGE)

assay

-

form

buffered aqueous glycerol solution

form

aqueous solution

form

buffered aqueous glycerol solution

form

-

recombinant

expressed in baculovirus infected Sf9 cells

recombinant

expressed in baculovirus infected Sf9 cells

recombinant

expressed in baculovirus infected Sf9 cells

recombinant

-

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

wet ice


Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters