Skip to Content
Merck

Skip To

F1127

D-Fructose 1-phosphate sodium salt

≥95%, amorphous powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C6H11O9PNa2
CAS Number:
Molecular Weight:
304.10
UNSPSC Code:
12352201
PubChem Substance ID:
MDL number:
eCl@ss:
38100201
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95%

form

amorphous powder

storage temp.

−20°C

SMILES string

[Na].OCC1OC(O)(COP(O)(O)=O)C(O)C1O



Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

WGK

WGK 3

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library