Skip to Content
Merck

Key Documents

Skip To

F8205

Sebomed basal medium

With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mL
Please contact Customer Service for Availability
1 550,00 CZK

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207

1 550,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

sterile-filtered, sterile

form

liquid

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

storage temp.

2-8°C

General description

Sebomed basal medium is ideal for controlled studies. It can be used to study the effects of hormones, cytokines, xenobiotics, and calcium on proliferation, differentiation, lipid synthesis, and cytokine production in the cell culture system. The basal medium is free of bovine products and does not contain phenol red or linoleic acid (or any other fatty acid).

Application

Sebomed basal medium has been used to culture immortalized human SZ95 sebocytes.


Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters