Skip to Content
Merck

Skip To

F9925

Anti-FLIPγ/δ, C-Terminal antibody produced in rabbit

1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CASH, Anti-CLARP, Anti-FLAME-1, Anti-I-FLICE

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IF, WB
Citations:
10


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 25 kDa (FLIPδ/FLIPS), antigen 35 kDa (FLIPγ)

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

immunocytochemistry: 2 μg/mL, immunofluorescence: 10 μg/mL, western blot: 1:1,000

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CFLAR(8837)
mouse ... Cflar(12633)

General description

In human Viral FLICE-inhibitory proteins (v-FLIPs) is identified as c-FLIP. It is composed of two death effector domains which have structural resemblance with the N-terminal half of caspase-8 and a caspase-like domain. It exist as multiple splice variants: FLIP α, β, γ and δ. Along with splice variants, it has two endogenous forms of the protein − c-FLIPlong and c-FLIPshort.

Immunogen

FLIP antibody was raised against a 19 amino acid peptide near the carboxy terminus of human FLIP.
The immunogen is located within amino acids 180 - 230 of FLIP.

Application

Anti-FLIPγ/δ, C-Terminal antibody is suitable for microarray and western blot at a dilution of 1:1,000 using HeLa, Jurkat, THP-1, A431, K562, and NIH-3T3 cell lysates.

Biochem/physiol Actions

C-FLIP plays an important role in apoptosis signaling pathways. It acts as proapoptotic molecule or as an anti-apoptotic molecule. It has been reported that c-FLIP can interact with both FADD and caspase-8. It prevents caspase-8 recruitment and processing through DED-DED (Death Effector Domain) interaction followed by CD95-induced apoptosis.

Physical form

Solution in 0.01 M phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters