Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C23H34O5
CAS Number:
Molecular Weight:
390.51
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
208-886-2
MDL number:
Beilstein/REAXYS Number:
96483
Pricing and availability is not currently available.
storage temp.
−20°C
SMILES string
[H][C@]12CC[C@]3([H])[C@]([H])(CC[C@]4(C)[C@H]([C@@H](O)C[C@]34O)C5=CC(=O)OC5)[C@@]1(C)CC[C@H](O)C2
Biochem/physiol Actions
Structure-activity relationships for 16β-substituted digitoxigenins as inhibitors of Na+/K+ ATPases.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
