Skip to Content
Merck

Skip To

GN655185

Greiner CELLSTAR® microplate

96 well, TC treated, flat bottom, suspension, chimney style, clear, sterile

Synonym(s):

96 well microplate, 96 well plate, cell culture plate, treated microplate, Greiner 655185

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41122107
Material:
flat bottom
Size:
96 wells
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

flat bottom

packaging

pack of 60 ea

manufacturer/tradename

Greiner 655185

availability

available only in China

technique(s)

cell culture: suitable

size

96 wells

suitability

suitable for (cell culture)

Legal Information

CELLSTAR is a registered trademark of Greiner Bio-One GmbH


Still not finding the right product?

Explore all of our products under Greiner CELLSTAR® microplate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library