Skip to Content
Merck

M0318

Anti-MRP3 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Multidrug Resistance Associated Protein 3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
MDL number:
eCl@ss:
32160702
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~180 kDa

species reactivity

human, rat

technique(s)

indirect immunofluorescence: 15-30 μg/mL using frozen sections of rat skin (outer root sheaths of hair follicles), western blot: 2.5-5 μg/mL using whole extract of cultured human colon carcinoma HCT-116 cells

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... ABCC3(8714)
mouse ... Abcc3(76408)
rat ... Abcc3(140668)

Immunogen

synthetic C-terminal peptide of human MRP3 (amino acids 1507-1527) with N-terminal cysteine conjugated to KLH. The sequence in mouse and rat differs by three residues.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library