Skip to Content
Merck

Skip To

M5437

Anti-MSK-1 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.44
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ARR, ICC, WB
Citations:
9


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 90 kDa

species reactivity

rat

technique(s)

immunocytochemistry: 1:1,000 using methanol/acetone-fixed rat fibroblast Rat-1 cell line, microarray: suitable, western blot: 1:2,000 using using whole cell lysate of rat fibroblast Rat-1 cell line

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPS6KA5(9252)
rat ... Rps6ka5(314384)

General description

The mitogen- and stress-activated kinases, MSK-1 and MSK-2 are activated by ERK/MAPK and p38 MAPK in response to growth and cell stress stimuli. MSK-1 and MSK-2 have 75% homology and interact with ERK1/2 and p38 through MAPK-binding domains near the C-terminus. There is a predominant expression of MSK proteins in heart, brain, skeletal muscles, lung, liver, kidney, pancreas and placenta. Highly conserved phosphorylation sites of MSK-1, Ser260 and Thr581, are followed by proline residues indicating that MSK-1 is activated by more than one MAPK. Endogenous MSK-1 is activated in cells by growth factors, phorbol ester stimulation, exposure to UV irradiation, and oxidative or chemical stress and is localized to nucleus of unstimulated or stimulated cells. MSK-1 is directly involved in the regulation of gene expression since it actively phosphorylates, stabilizes and activates many transcription factors. The major transcription factors that are regulated by MSK-1 include CREB, NF-κB and STAT3. MSKs also evidently phosphorylate Histone H3 at Ser10 and Ser28 and HMG at Ser6. Thus a primary role of MSK proteins is also promotion of gene relaxation and activation by regulating the binding of transcription repressors. Anti-MSK-1 specifically recognizes rat MSK-1 (90 kDa).

Immunogen

synthetic peptide corresponding to the C-terminus of human MSK1 (amino acids 783-800). This sequence is not found in the human and mouse MSK2 isoforms.

Application

Anti-MSK-1 may be used at a minimum working dilution of 1:2000 (immunoblotting in whole cell extracts) or 1:1000 (immunocytochemistry) in fibroblast Rat1 cell line. The antobody has also been used for detection by immunoblotting in mouse fibroblast 10T1/2 cells.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

— or —

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters