Skip to Content
Merck

Skip To

MAK613

Leucine Aminopeptidase Activity Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

LAP Activity Assay Kit

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41116104
eCl@ss:
40050204
Sign in to see your pricing
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 100 colorimetric tests

detection method

colorimetric

storage temp.

−20°C

General description

Leucine aminopeptidase (LAP, EC 3.4.11.1) removes N-terminalleucine residues from peptides and proteins and is abundant in liver and intestinal tissues. LAP activity can increase in settings of tissue damage, tumorigenesis, or inflammation.

Application

For quantitative determination of LAP enzyme activity in biologicalsamples, such as cells, tissues, and serum samples.


For high-throughput screening of LAP inhibitors and evaluation of drug modulators.

Biochem/physiol Actions

The Leucine Aminopeptidase Activity Assay Kit provides a rapid colorimetric method in which LAP hydrolyzes a dye-linked leucine substrate to yield a yellow product measurable at 405 nm. The change in absorbance over 20 minutes is proportional to LAP activity. The linear detection range in 96-well plate is 0.025 to 250 U/L.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MAK120MAK501
description

sufficient for 100 colorimetric tests

description

sufficient for 100 colorimetric tests

description

sufficient for 100 colorimetric or fluorometric tests

usage

sufficient for 100 colorimetric tests

usage

sufficient for 100 colorimetric tests

usage

-

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric, fluorometric

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C


Storage Class

10 - Combustible liquids

WGK

nwg

Flash Point (°F)

188.6 °F - closed cup

Flash Point (°C)

87 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library