Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Please contact Customer Service for Availability | 5 940,00 CZK |
About This Item
NACRES:
NA.24
UNSPSC Code:
23201100
5 940,00 CZK
Please contact Customer Service for Availability
Quality Segment
assay
≥95% (HPLC)
form
lyophilized
technique(s)
protein mass spectrometry: suitable
color
off-white
shipped in
ambient
storage temp.
−20°C
Analysis Note
General Description
SILu™Pep Oxyntomodulin is a synthetic, stable isotope-labeled human peptide.
Sequence
HSQGTFTSDYSKY-Leu(U13C6)-DSRRAQDFVQW-Leu(U13C6)-MNTKRNRNNIA
Terminal modifications: N-Term: [H]
Length (residues): 30
Label Incorporation
>= 98% as determined by mass spectrometry
SILu™Pep Oxyntomodulin is a synthetic, stable isotope-labeled human peptide.
Sequence
HSQGTFTSDYSKY-Leu(U13C6)-DSRRAQDFVQW-Leu(U13C6)-MNTKRNRNNIA
Terminal modifications: N-Term: [H]
Length (residues): 30
Label Incorporation
>= 98% as determined by mass spectrometry
Legal Information
SILu is a trademark of Sigma-Aldrich Co. LLC
1 of 1
This Item | |||
|---|---|---|---|
| description synthetic, SIL MS Peptide Standard, ¹³C labeled | description synthetic, SIL MS Peptide Standard, ¹³C labeled | description synthetic, SIL MS Peptide Standard, ¹³C labeled | description synthetic, SIL MS Peptide Standard, ¹³C labeled |
| assay ≥95% (HPLC) | assay ≥95% (HPLC) | assay ≥95% (HPLC) | assay ≥95% (HPLC) |
| technique(s) protein mass spectrometry: suitable | technique(s) protein mass spectrometry: suitable | technique(s) protein mass spectrometry: suitable | technique(s) protein mass spectrometry: suitable |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| form lyophilized | form lyophilized | form lyophilized | form lyophilized |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| color off-white | color off-white | color off-white | color off-white |
Storage Class
11 - Combustible Solids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



