Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
Please contact Customer Service for Availability | 6 380,00 CZK |
About This Item
6 380,00 CZK
material
red (with black Biohazard warning)
packaging
pack of 1 ea
manufacturer/tradename
MTC™ Bio A8002B
W × L
19 in. (48 cm) × 19 in. (48 cm)
capacity
26 gal (100 L)
General description
Features and Benefits
- International biohazard symbol
- Foot pedal for hands free operation and reduced risk of contamination
- Simple bag installation
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description capacity 26 gal (100 L), W × L 19 in. (48 cm) × 19 in. (48 cm) | description capacity 30 L (50 gal), W × L × H 6 in. (41 cm) × 16 in. (41 cm) × 24 in. (60 cm), pack of 1 ea | description W × L × H 10 in. (25 cm) × 14 in. (35 cm) × 19 in. (48 cm), capacity 8 gal (30 L) | description high-density polyethylene, capacity 33 gal (113 L), W × L 33 in. (84 cm) × 41 in. (104 cm), pack of 100 ea |
| manufacturer/tradename MTC™ Bio A8002B | manufacturer/tradename MTC™ Bio A8006B | manufacturer/tradename MTC™ Bio A8000B | manufacturer/tradename MTC™ Bio A8003R |
| material red (with black Biohazard warning) | material red (with black Biohazard warning) | material red (with black Biohazard warning) | material red , high-density polyethylene |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 100 ea |
| capacity 26 gal (100 L) | capacity 30 L (50 gal) | capacity 8 gal (30 L) | capacity 33 gal (113 L) |
| W × L 19 in. (48 cm) × 19 in. (48 cm) | W × L - | W × L - | W × L 33 in. (84 cm) × 41 in. (104 cm) |
Still not finding the right product?
Explore all of our products under MTC™ Bio BowTie™ Floor standing Biohazard Bin With Lid
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



