Skip to Content
Merck

Skip To

MTCA8006B

MTC™ Bio BowTie™ Floor standing Biohazard Bin With Lid

capacity 30 L (50 gal), W × L × H 6 in. (41 cm) × 16 in. (41 cm) × 24 in. (60 cm), pack of 1 ea

Synonym(s):

MTC™ Bio BowTie™ Floor standing Biohazard Bin With Lid, bio waste bin, MTC Bio A8006B, lab waste bag, waste disposal bag

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

Available to ship TODAYfromSchnelldorf Distribution

3 570,00 CZK

About This Item

UNSPSC Code:
24111500
NACRES:
NB.43
Manufacturer/tradename:
MTC™ Bio A8006B
Material:
red (with black Biohazard warning)
Packaging:
pack of 1 ea

3 570,00 CZK


Available to ship TODAYDetails


​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

red (with black Biohazard warning)

packaging

pack of 1 ea

manufacturer/tradename

MTC™ Bio A8006B

W × L × H

6 in. (41 cm) × 16 in. (41 cm) × 24 in. (60 cm)

capacity

30 L (50 gal)

General description

Biohazard bins perfect for all of the infectious orcontaminated waste produced in laboratories on a daily basis. Clearly marked with the international biohazard symbol, there is no need for additionalstickers or labels. The foot-pedal operated allows for hand free operation and reduces contamination risks. Notch hooks on the outside of the basket rim makeinstalling bags simple and easy. For use with the appropriate sized BowTie™ bag.

Features and Benefits

  • International biohazard symbol
  • Foot pedal for hands free operation and reduced risk of contamination
  • Simple bag installation

Legal Information

BowTie is a trademark of MTC Bio
MTC is a trademark of MTC Bio

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MTCA8002BMTCA8000BMTCA8003R
description

capacity 30 L (50 gal), W × L × H 6 in. (41 cm) × 16 in. (41 cm) × 24 in. (60 cm), pack of 1 ea

description

capacity 26 gal (100 L), W × L 19 in. (48 cm) × 19 in. (48 cm)

description

W × L × H 10 in. (25 cm) × 14 in. (35 cm) × 19 in. (48 cm), capacity 8 gal (30 L)

description

high-density polyethylene, capacity 33 gal (113 L), W × L 33 in. (84 cm) × 41 in. (104 cm), pack of 100 ea

manufacturer/tradename

MTC™ Bio A8006B

manufacturer/tradename

MTC™ Bio A8002B

manufacturer/tradename

MTC™ Bio A8000B

manufacturer/tradename

MTC™ Bio A8003R

material

red (with black Biohazard warning)

material

red (with black Biohazard warning)

material

red (with black Biohazard warning)

material

red , high-density polyethylene

packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 100 ea

W × L × H

6 in. (41 cm) × 16 in. (41 cm) × 24 in. (60 cm)

W × L × H

-

W × L × H

10 in. (25 cm) × 14 in. (35 cm) × 19 in. (48 cm)

W × L × H

-

capacity

30 L (50 gal)

capacity

26 gal (100 L)

capacity

8 gal (30 L)

capacity

33 gal (113 L)


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library