Skip to Content
Merck

Skip To

MTCD309605

MTC™ Bio OptiWell™ Deep Well Plate

96 round wells, flat bottom wells, capacity 0.4 mL, non-sterile, case of 100 ea (10 bags of 10ea)

Synonym(s):

automated research plate, MTC Bio D3096-05, automated assay plate, lab automation plate, research automation tray, robotic research plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
Please contact Customer Service for Availability
14 200,00 CZK

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
Material:
clear plate, clear wells, flat bottom wells, polypropylene , round wells
Size:
96 wells
Sterility:
non-sterile
Feature:
lid: no, skirt: no

14 200,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

clear plate, clear wells, flat bottom wells, polypropylene , round wells

sterility

non-sterile

feature

lid: no, skirt: no

packaging

case of 100 ea (10 bags of 10ea)

manufacturer/tradename

MTC™ Bio D3096-05

parameter

4,000 × g speed

capacity

0.4 mL

size

96 wells

well volume

0.4 mL

General description

OptiWell plates come in 24, 48, 96 or 384 format for virtually every research lab, HTS application and automated/robotic platform. These plates are useful for bacterial cultures, centrifugation (up to 4000 x g) and general sample storage. Plates are produced/packaged in a Class 100,000 clean room and are ANSI / SLAS 1-2004 compliant. Several plates are sterilized by gamma irradiation and all are certified non-pyrogenic, RNase/DNase free.

Features and Benefits

  • For automated research platforms / robotic equipment
  • Produced/packaged in a Class 100,000 clean room
  • ANSI / SLAS 1-2004 compliant
  • Minimal binding / liquid retention
  • Black printed alpha-numerics
  • Virgin, medical-grade polypropylene
  • Sterilized by gamma irradiation
  • Certified non-pyrogenic, RNase/DNase free

Legal Information

MTC is a trademark of MTC Bio
OptiWell is a trademark of MTC Bio

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MTCD309603MTCD309609MTCD309611
description

96 round wells, flat bottom wells, capacity 0.4 mL, non-sterile, case of 100 ea (10 bags of 10ea)

description

96 round wells, U-bottom wells, capacity 0.4 mL, non-sterile, case of 100 ea (10 bags of 10ea)

description

96 square wells, V-bottom wells, capacity 0.5 mL, non-sterile, case of 100 ea (10 bags of 10ea)

description

96 round wells, U-bottom wells, capacity 0.5 mL, non-sterile, case of 100 ea (10 bags of 10ea)

feature

skirt: no, lid: no

feature

skirt: no, lid: no

feature

skirt: no, lid: no

feature

lid: no, skirt: no

manufacturer/tradename

MTC™ Bio D3096-05

manufacturer/tradename

MTC™ Bio D3096-03

manufacturer/tradename

MTC™ Bio D3096-09

manufacturer/tradename

MTC™ Bio D3096-11

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

material

clear plate, clear wells, polypropylene , flat bottom wells, round wells

material

clear plate, polypropylene , clear wells, round wells, U-bottom wells

material

clear wells, clear plate, square wells, V-bottom wells, polypropylene

material

clear wells, U-bottom wells, clear plate, round wells, polypropylene

size

96 wells

size

96 wells

size

96 wells

size

96 wells

packaging

case of 100 ea (10 bags of 10ea)

packaging

case of 100 ea (10 bags of 10ea)

packaging

case of 100 ea (10 bags of 10ea)

packaging

case of 100 ea (10 bags of 10ea)


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library