Merck
MTCD309633S
MTC™ Bio OptiWell™ Deep Well Plate
96 square wells, V-bottom wells, capacity 1.6 mL, sterile, case of 50 ea (10 bags of 5ea)
Synonym(s):
automated research plate, MTC Bio D3096-33S, automated assay plate, lab automation plate, research automation tray, robotic research plate
Select a Size
Change View
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
eCl@ss:
32150313
Material:
V-bottom wells, clear plate, clear wells, polypropylene , square wells
Size:
96 wells
Sterility:
sterile
Feature:
lid: no, skirt: no
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
V-bottom wells, clear plate, clear wells, polypropylene , square wells
sterility
sterile
feature
lid: no, skirt: no
packaging
case of 50 ea (10 bags of 5ea)
manufacturer/tradename
MTC™ Bio D3096-33S
parameter
4,000 × g speed
capacity
1.6 mL
size
96 wells
well volume
1.6 mL
General description
OptiWell plates come in 24, 48, 96 or 384 format for virtually every research lab, HTS application and automated/robotic platform. These plates are useful for bacterial cultures, centrifugation (up to 4000 x g) and general sample storage. Plates are produced/packaged in a Class 100,000 clean room and are ANSI / SLAS 1-2004 compliant. Several plates are sterilized by gamma irradiation and all are certified non-pyrogenic, RNase/DNase free.
Features and Benefits
  • For automated research platforms / robotic equipment
  • Produced/packaged in a Class 100,000 clean room
  • ANSI / SLAS 1-2004 compliant
  • Minimal binding / liquid retention
  • Black printed alpha-numerics
  • Virgin, medical-grade polypropylene
  • Sterilized by gamma irradiation
  • Certified non-pyrogenic, RNase/DNase free
Legal Information
MTC is a trademark of MTC Bio
OptiWell is a trademark of MTC Bio
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)