Skip to Content
Merck

MTCD33205S

MTC™ Bio OptiWell Silicone Plate Sealing Mat

size 96 wells, square wells, + opening, well diam. 8.4 mm, sterile, pack of 50 ea

Synonym(s):

plate cover, rubber plate seal, MTC Bio D3320-5S, plate sealing pad, silicone plate cover, silicone plate seal

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122410
eCl@ss:
32120613
Manufacturer/tradename:
MTC™ Bio D3320-5S
Material:
clear , silicone , square wells
Sterility:
sterile
Packaging:
pack of 50 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

clear , silicone , square wells

sterility

sterile

feature

autoclavable

packaging

pack of 50 ea

manufacturer/tradename

MTC™ Bio D3320-5S

parameter

-40-80 °C temp. range

size

96 wells

well diam.

8.4 mm

General description

OptiWell Sealing Mats are produced from high quality, chemically resistant silicone and are designed to provide an airtight and liquid-tight seal. The material resists tearing and wear, minimizing the potential for particulate debris. They are compatible with temperatures from -40 to 80°C, can be autoclaved and are also prepped for reuse with a 10% bleach wash with an ethanol rinse. They can also feature black alphanumeric printing for easy reference and two types of resealable perforations ("+" and "-"), RNase, DNase and pyrogen free.

Features and Benefits

  • Black alphanumeric coding
  • Forms a tight seal in plate wells
  • Perforated mats reseal themselves
  • RNase, DNase and pyrogen free

Legal Information

MTC is a trademark of MTC Bio


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library