Skip to Content
Merck

Skip To

MTCD339012

MTC™ Bio OptiWell Automation Reservoir

capacity 180 mL, 12 channel, polypropylene, non-sterile, case of 50 ea (10 bags of 5ea)

Synonym(s):

liquid reservoir, MTC Bio D3390-12, liquid receptacle, reagent dish, reagent holder, reservoir

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
Please contact Customer Service for Availability
8 040,00 CZK

About This Item

UNSPSC Code:
41121522
Manufacturer/tradename:
MTC™ Bio D3390-12
Capacity:
180 mL
Material:
clear , polypropylene
Sterility:
non-sterile

8 040,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

clear , polypropylene

sterility

non-sterile

feature

12 channel, autoclavable

packaging

case of 50 ea (10 bags of 5ea)

manufacturer/tradename

MTC™ Bio D3390-12

parameter

-80 °C temp. range

H

31.5 mm

capacity

180 mL

General description

OptiWell Reservoirs are compatible with a wide range of automated liquid handling equipment. Offered with 1 to 384 wells, they are useful in multiple applications, including mixing, fraction collection, plate replication, compound storage, and scale-up / scale-down methods. OptiWell Reservoirs conform with SLAS standard dimensions. They are made of chemically inert, low binding polypropylene to minimize excess residue/waste.

Features and Benefits

  • ANSI / SLAS 1-2004 Compliant
  • Certified RNase, DNase and pyrogen free
  • Robot friendly / standard dimensions
  • Autoclavable and freezable to -80°C
  • SLAS standard footprint

Legal Information

MTC is a trademark of MTC Bio

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MTCD33908MTCD339038MTCD339012S
description

capacity 180 mL, 12 channel, polypropylene, non-sterile, case of 50 ea (10 bags of 5ea)

description

capacity 180 mL, 8 channel, polypropylene, non-sterile, case of 50 ea (10 bags of 5ea)

description

capacity 185 mL, 384 channel, polypropylene, non-sterile, case of 50 ea (10 bags of 5ea)

description

capacity 180 mL, 12 channel, polypropylene, sterile, case of 50 ea (10 bags of 5ea)

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

sterile

capacity

180 mL

capacity

180 mL

capacity

185 mL

capacity

180 mL

manufacturer/tradename

MTC™ Bio D3390-12

manufacturer/tradename

MTC™ Bio D3390-8

manufacturer/tradename

MTC™ Bio D3390-38

manufacturer/tradename

MTC™ Bio D3390-12S

material

clear , polypropylene

material

clear , polypropylene

material

polypropylene , clear

material

polypropylene , clear

packaging

case of 50 ea (10 bags of 5ea)

packaging

case of 50 ea (10 bags of 5ea)

packaging

case of 50 ea (10 bags of 5ea)

packaging

case of 50 ea (10 bags of 5ea)

feature

12 channel, autoclavable

feature

8 channel, autoclavable

feature

autoclavable, 384 channel

feature

12 channel, autoclavable


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library