Merck
Skip To
MTCM4035
MTC™ Bio Inoculating Loops and Needles
flexible loop, capacity 5 μL, polypropylene, sterile, case of 1000 ea (50 peel packs of 20ea)
Synonym(s):
culture needle, cutlure loop, MTC Bio M4035, sampling loop, sampling tool
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
Please contact Customer Service for Availability
2 680,00 CZK
About This Item
UNSPSC Code:
41161502
2 680,00 CZK
Please contact Customer Service for Availability
​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polypropylene
sterility
sterile
packaging
case of 1000 ea (50 peel packs of 20ea)
manufacturer/tradename
MTC™ Bio M4035
capacity
5 μL
General description
Sterile disposable plastic loops and needles designed for use in quantitative procedures such as sampling, urine counts, and serial dilutions, as well as for bacterial inoculation. Because they are pre-sterilized and designed for single use, there is no risk of cross-contamination and no need for flaming. The unique hexagonal shape of the shaft makes it easier to grip, orientate, and guide through a smooth streaking process. The loop surfaces are extremely smooth with no rough edges and no flashing or burrs on the loop head. This results in the elimination of cutting or gouging of the agar surface.
Flexible Polypropylene Loops - Make it easier to maneuver them within Petri dishes, test tubes, or other confined spaces, improving the researcher′s ability to collect and transfer samples efficiently. The flexibility allows for more precise sampling from cultures, especially when working with small or delicate colonies that could be easily damaged by more rigid instruments.
Rigid Polystyrene Loops & Needles - Allow for more precise streaking patterns on agar plates, which is essential for isolating individual colonies or performing quantitative assays. They are also better suited for handling viscous or dense samples, where flexibility might impede the ability to effectively collect and transfer the sample.
Features and Benefits
  • Sterilized to SAL 10-6
  • Easy-grip hexagonal handle design
  • Flexible PP or Rigid PS available
  • Individually wrapped or peel packs of 20
Legal Information
MTC is a trademark of MTC Bio
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
MTCM4035WMTCM4017MTCM4017W
description
flexible loop, capacity 5 μL, polypropylene, sterile, case of 1000 ea (50 peel packs of 20ea)
description
flexible loop, capacity 5 μL, polypropylene, sterile, case of 1000 ea (individually wrapped)
description
rigid loop, capacity 1 μL, polystyrene, sterile, case of 1000 ea (50 peel packs of 20ea)
description
rigid loop, capacity 1 μL, polystyrene, sterile, case of 1000 ea (individually wrapped)
material
polypropylene
material
polypropylene
material
polystyrene
material
polystyrene
capacity
5 μL
capacity
5 μL
capacity
1 μL
capacity
1 μL
manufacturer/tradename
MTC™ Bio M4035
manufacturer/tradename
MTC™ Bio M4035-W
manufacturer/tradename
MTC™ Bio M4017
manufacturer/tradename
MTC™ Bio M4017-W
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
packaging
case of 1000 ea (50 peel packs of 20ea)
packaging
case of 1000 ea (individually wrapped)
packaging
case of 1000 ea (50 peel packs of 20ea)
packaging
case of 1000 ea (individually wrapped)
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)