Skip to Content

OGS140

PSF-CMV-COOH-EKT-GST3 - C-TERMINAL GST PLASMID

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.85
UNSPSC Code:
12352200
Promoter:
Promoter name: CMV
Promoter activity: constitutive
Promoter type: mammalian
Origin of replication:
pUC (500 copies)
Bacteria selection:
kanamycin
Reporter gene:
none
Peptide cleavage:
EKT
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


tag

GST tagged

form

buffered aqueous solution

mol wt

size 4900 bp

bacteria selection

kanamycin

origin of replication

pUC (500 copies)

peptide cleavage

EKT

peptide tag location

C-terminal

promoter

Promoter name: CMV
Promoter activity: constitutive
Promoter type: mammalian

reporter gene

none

shipped in

ambient

storage temp.

−20°C

General description

This vector adds a GST tag positioned at the 3 prime end (C-terminus) of a gene (expressed protein) that is within the primary MCS (NotI to XbaI). The tag can be used to purify a protein by binding to glutathione and the tag can also be used to increase the solubility of recombinant protein produced in bacterial cells. For reference the coding sequence of the GST tag is one base pair out of frame with the TAG stop codon that is found within the upstream XbaI site. There is an enterokinase cleavage site immediately upstream of this tag (DDDDK) that can be used to remove the tag from a purified protein if required.

Promoter Expression Level: This plasmid contains the mammalian CMV promoter to drive gene expression. We have tested all of our mammalian promoters in a range of cell types and CMV is consistently the strongest in those we have studied. However there are many reports of the CMV promoter demonstrating silencing by methylation in long-term culture.

Application

Cloning in a gene: There is a start codon in the NcoI site can be removed by digestion with KpnI if required. The MCS for gene insertions extends from NotI to XbaI. There are built in Shine-Dalgarno and KOZAK sequences aligned with the start codon within the NcoI restriction site. If these are removed you will need to add a ribosomal entry site to the 5 prime end of your gene for optimal expression.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxfordgenetics.com.

Other Notes

Looking for more vector options to move your experiments forward faster? Consider a custom cloning vector designed and built by Oxford Genetics. Find out more at Oxford Genetics - Sigma's partner for cloning and expression vectors for molecular biology and synthetic biology applications.
Please select the file type you require. For reference most cloning programs will import a .gb (Genbank) file and will show all of the plasmids features automatically when downloaded and imported.

Genebank Vector Sequence File

FASTA Vector Sequence File

Full Plasmid Map

Legal Information

Oxford Genetics is a trademark of Oxford Genetics Ltd


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library