Sign In to View Organizational & Contract Pricing.
About This Item
NACRES:
NA.85
UNSPSC Code:
12352200
Promoter:
Promoter name: TEF1
Promoter activity: constitutive
Promoter type: yeast
Promoter activity: constitutive
Promoter type: yeast
Origin of replication:
2Micron, pUC (500 copies)
Bacteria selection:
kanamycin
Reporter gene:
none
Peptide cleavage:
Fxa
conjugate
streptavidin conjugate
form
buffered aqueous solution
mol wt
size 6753 bp
bacteria selection
kanamycin
origin of replication
2Micron, pUC (500 copies)
peptide cleavage
Fxa
peptide tag location
C-terminal
promoter
Promoter name: TEF1
Promoter activity: constitutive
Promoter type: yeast
reporter gene
none
shipped in
ambient
storage temp.
−20°C
yeast selection
uracil
General description
This plasmid is designed to express tagged proteins in yeast cells (Saccharomyces cerevisiae). The plasmid contains an auxotrophic Uracil selection expression cassette (URA3) that allows for the positive selection of yeast that are deficient in the URA3 gene (YEL021W). This is typically achieved by growing the yeast in minimal media that is reconstituted with the essential amino acids and nucleotides but excluding Uracil.
About the Peptide Tag:This plasmid contains a c-terminal Strep affinity tag that can be fused to a gene of interest to allow protein detection and/or purification. The sequence of the tag is: WSHPQFEK.
About the Cleavage Tag:This plasmid also encodes a protease cleavage site that is designed to be positioned between your gene of interest and the tag to allow the removal of the tag following protein purification or isolation. This plasmid contains a FXa cleavage tag. The protein sequence of the cleavage tag is: IEGR. It cleaves after the Arg residue but can also cleave less frequently at secondary basic sites. Its most common secondary cleave site is between the Gly and Arg residues although the frequency of these events is protein specific.
Promoter Expression Level: This plasmid contains the Yeast Elongation Factor Alpha promoter (TEF1). This is the strongest of the yeast promoters that we sell. It is a constitutive promoter and requires no induction. If you are interested in weaker promoters levels than we also stock plasmids that contain the following promoters in order of decreasing strength TPI (strong), ADHI (medium), STE5 (weak). We also stock Galactose inducible promoter plasmids if inducible expression is required. Please contact us for further information.
About the Peptide Tag:This plasmid contains a c-terminal Strep affinity tag that can be fused to a gene of interest to allow protein detection and/or purification. The sequence of the tag is: WSHPQFEK.
About the Cleavage Tag:This plasmid also encodes a protease cleavage site that is designed to be positioned between your gene of interest and the tag to allow the removal of the tag following protein purification or isolation. This plasmid contains a FXa cleavage tag. The protein sequence of the cleavage tag is: IEGR. It cleaves after the Arg residue but can also cleave less frequently at secondary basic sites. Its most common secondary cleave site is between the Gly and Arg residues although the frequency of these events is protein specific.
Promoter Expression Level: This plasmid contains the Yeast Elongation Factor Alpha promoter (TEF1). This is the strongest of the yeast promoters that we sell. It is a constitutive promoter and requires no induction. If you are interested in weaker promoters levels than we also stock plasmids that contain the following promoters in order of decreasing strength TPI (strong), ADHI (medium), STE5 (weak). We also stock Galactose inducible promoter plasmids if inducible expression is required. Please contact us for further information.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.oxgene.com
Other Notes
To view sequence information for this product, please visit the product page
Storage Class
12 - Non Combustible Liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.