Skip to Content
Merck

Skip To

OGS211

PSF-CMV-NH2-HIS-EKT-NCOI - N-TERMINAL HIS TAG PLASMID

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.85
UNSPSC Code:
12352200
Promoter:
Promoter name: CMV
Promoter activity: constitutive
Promoter type: mammalian
Origin of replication:
pUC (500 copies)
Bacteria selection:
kanamycin
Reporter gene:
none
Peptide cleavage:
EKT


tag

6-His tagged

form

buffered aqueous solution

mol wt

size 6152 bp

bacteria selection

kanamycin

origin of replication

pUC (500 copies)

peptide cleavage

EKT

peptide tag location

N-terminal

promoter

Promoter name: CMV
Promoter activity: constitutive
Promoter type: mammalian

reporter gene

none

shipped in

ambient

storage temp.

−20°C

General description

This vector adds a His epitope tag to the N-terminus of a protein that is encoded within the multiple cloning site. This tag allows the purification of a tagged protein by binding to metal matrices such as nickel or cobalt. There is an enterokinase cleavage site (DDDDK) immediately downstream of the His tag that can be used to remove the His tag from a purified protein. It cleaves after the lysine residue.

Promoter Expression Level: This plasmid contains the mammalian CMV promoter to drive gene expression. We have tested all of our mammalian promoters in a range of cell types and CMV is consistently the strongest in those we have studied. However there are many reports of the CMV promoter demonstrating silencing by methylation in long-term culture.

Application

Multiple cloning site notes: The his tag is in frame with the ATG start codon that is within the NcoI restriction site allowing fusion with any genes that we sell in the MCS that contain an NcoI at the 5 prime. This tag is positioned immediately upstream of the NcoI site to minimise the amount of extra amino acids that are added to the N-terminus of your gene.

The start codon that is within the NcoI restriction site is in frame with and adjacent to the coding sequence for the His/EKT peptide tag. Shine-Dalgarno sequences and KOZAK sequences are positioned correctly at the start of the His signal peptide. The ClaI to NheI sites have other functions such as adding peptide tags or IRES elements. The BsgI and BseRI restriction sites cleave within the stop codon in the XbaI site and allow fusion with our C-terminal peptide tags provided that your genes stop codon is also positioned here.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxgene.com

Other Notes

To view sequence information for this product, please visit the product page


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters