Sign In to View Organizational & Contract Pricing.
About This Item
NACRES:
NA.85
UNSPSC Code:
12352200
Promoter:
Promoter name: OXB20
Promoter activity: constitutive
Promoter type: bacterial
Promoter activity: constitutive
Promoter type: bacterial
Origin of replication:
pUC (500 copies)
Bacteria selection:
kanamycin
Reporter gene:
none
Peptide cleavage:
no cleavage
recombinant
expressed in E. coli
tag
6-His tagged
form
buffered aqueous solution
mol wt
size 3929 bp
technique(s)
molecular cloning: suitable
bacteria selection
kanamycin
origin of replication
pUC (500 copies)
peptide cleavage
no cleavage
peptide tag location
N-terminal
promoter
Promoter name: OXB20
Promoter activity: constitutive
Promoter type: bacterial
reporter gene
none
secretion signal
OmpA
shipped in
ambient
storage temp.
−20°C
General description
This plasmid is designed to express tagged proteins in E. coli. The plasmid contains a constitutive promoter (OXB20) derived from the region upstream of the E. coli RecA gene. It does not require induction or any additional components for activity. It is the strongest of the bacterial promoters that we provide and this high level of expression can cause expression problems with some proteins with poor solubility. For this reason we sell a range of bacterial promoters with different expression levels (OXB1(low)>OXB20(high)) that can be provided with the peptide tags in this plasmid on request.
About the Peptide Tag:This plasmid contains an Outer Membrane Protein A (OmpA) secretory signal peptide (SP) to allow proteins to be exported from the cytosol. During translocation from the cytosol the signal peptide is removed from the protein by endogenous proteases. This plasmid also contains a secondary Hexa-Histidine (6His) protein tag. The sequence of this tag is: HHHHHHWe provide a range of dual peptide tag plasmids. This is because some peptide tags provide specific biological properties (e. g., small molecule affinity new epitopes solubility or protein secretion) that are not provided by others.
Promoter Expression Level: This plasmid contains a constitutive bacterial promoter that does not require induction. It is the strongest bacterial promoter we sell and this can cause solubility and expression problems with some proteins. We also offer a range of other bacterial promoters that are compatible with this plasmid and are available on request.
About the Peptide Tag:This plasmid contains an Outer Membrane Protein A (OmpA) secretory signal peptide (SP) to allow proteins to be exported from the cytosol. During translocation from the cytosol the signal peptide is removed from the protein by endogenous proteases. This plasmid also contains a secondary Hexa-Histidine (6His) protein tag. The sequence of this tag is: HHHHHHWe provide a range of dual peptide tag plasmids. This is because some peptide tags provide specific biological properties (e. g., small molecule affinity new epitopes solubility or protein secretion) that are not provided by others.
Promoter Expression Level: This plasmid contains a constitutive bacterial promoter that does not require induction. It is the strongest bacterial promoter we sell and this can cause solubility and expression problems with some proteins. We also offer a range of other bacterial promoters that are compatible with this plasmid and are available on request.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.oxgene.com
Other Notes
To view sequence information for this product, please visit the product page
Storage Class
12 - Non Combustible Liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.