Skip to Content
Merck

Skip To

OGS52

PSF-H1 - SIRNA/RNAI EXPRESSION PLASMID

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.85
UNSPSC Code:
12352200
eCl@ss:
32160703
Promoter:
Promoter name: H1
Promoter activity: siRNA
Promoter type: mammalian
Origin of replication:
pUC (500 copies)
Bacteria selection:
kanamycin
Reporter gene:
none
Peptide cleavage:
no cleavage


form

buffered aqueous solution

mol wt

size 3801 bp

technique(s)

molecular cloning: suitable

bacteria selection

kanamycin

origin of replication

pUC (500 copies)

peptide cleavage

no cleavage

promoter

Promoter name: H1
Promoter activity: siRNA
Promoter type: mammalian

reporter gene

none

shipped in

ambient

storage temp.

−20°C

General description

The expression of short hairpin RNA in eukaryotic cells. The H1 is transcribed by RNA polymerase III (RNA pol III) which terminates transcription at a string of thymidine residues typically a minimum of five residues in mammalian cells. Unlike most SnapFastTM vectors this plasmid contains an RNA pol III terminator downstream of the XhoI site although it is more common to insert a terminator sequence at the end of the sequence you insert into the MCS that encodes a shRNA.

Promoter Expression Level:

Application

Multiple cloning site notes: This vector has a modified MCS compared to standard plasmids. The ATG start codon in the NcoI site has been removed as have the Shine-Dalgarno and KOZAK sequences as these are not required for RNA Pol3 expressed transcripts.

The terminator for RNA polymerase III has been positioned immediately downstream of the XhoI site. There is an additional string of Thymidine residues (the terminator sequence for RNA Pol III) at the end of the SV40 polyA region however using this site will produce a long transcript and has not been validated. We recommend inserting your hairpin DNA sequence between Hind3 and XhoI. The theoretical RNA position +1 is the first base of the NotI restriction site.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxgene.com

Other Notes

To view sequence information for this product, please visit the product page


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters