Skip to Content
Merck

Skip To

P247

Monoclonal Anti-phospho-CaM Kinase IIα (pThr286) antibody produced in mouse

clone 22B1, purified immunoglobulin, buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Please contact Customer Service for Availability
16 900,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.44
MDL number:
Conjugate:
unconjugated
Clone:
22B1, monoclonal
Application:
IF, WB
Citations:
5

16 900,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

22B1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~50 kDa

species reactivity

rat, mouse

technique(s)

indirect immunofluorescence: 1:100 using rat hippocampus (cold methanol fixation), western blot: 1:2,000

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr286)

Gene Information

mouse ... Camk2a(12322)
rat ... Camk2a(25400)

General description

Intracellular Calcium (Ca2+) acts as major secondary messenger that mediates signalling induced by hormones and neurotransmitters. Cellular responses to calcium are modulated by a family of Ca2+/Calmodulin dependent protein kinases. The members of this family include CaMK I, CAMK II and CAMK IV. These kinases mediate rapid, highly specific and reversible post-translational modification involving ATP. CaMK II is abundantly expressed in mammalian brain, with highest expression in neurons of hippocampus and cerebral cortex, and plays critical role in synaptic transmission, memory, learning and behaviour. CaMK II family has four members, CaMK IIα, CaMK IIβ, CaMK IIγ and CaMK IIδ. The CaMK IIα?and β?isoforms are predominantly expressed in the brain, localized mainly in the cytosol and postsynaptic densities (PSDs), whereas the CaMK IIγ and δ isoforms are expressed in all tissues. CaMK II is autophosphorylated at α-Thr286 residue in response to changes in the concentration of intracellular calcium. This may prolong physiological responses to temporary changes in Ca2+ concentrations. CaMK II regulates synaptic signalling by phosphorylating glutamate receptors in the neurons. A distinct CaMK II has also been implicated in the process of smooth muscle contraction by phosphorylation of myosin light chain kinase.
This product reacts with the recombinant and native rat phosphorylated CaMK II α subunit only (50 kDa). It does not cross-react with the non-phosphorylated α subunit or with the 60 kDa β subunit in either phosphorylation state.

Immunogen

phosphorylated (at Thr286) synthetic phosphopeptide corresponding to amino acids of rat and mouse CaM kinase IIα subunit that contains threonine 286.

Application

Anti-phospho-CaM Kinase IIα (pThr286) antibody may be used in indirect immunofluorescence at a working dilution of 1:100 in rat hippocampal cells fixed with cold methanol. Detection by immunoblotting is possible at a working dilution of 1:1000.

Biochem/physiol Actions

The antibody reacts specifically with both the recombinant and native rat phosphorylated CaM (Ca2+/calmodulin) dependent kinase IIα subunit. It does not cross-react with the non-phosphorylated α subunit or with the β subunit (60 kDa) in either phosphorylation state. By immunofluorescence on rat hippocampal cells, the antibody labels cell somas and neuropil areas, while nuclei are only lightly stained.

Physical form

Solution in phosphate buffered saline containing 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C265SAB4504356SAB4301224
description

clone 22B1, purified immunoglobulin, buffered aqueous solution

description

clone 6G9, purified immunoglobulin, buffered aqueous solution

description

affinity isolated antibody

description

affinity isolated antibody

species reactivity

rat, mouse

species reactivity

rat, mouse

species reactivity

rat, human, mouse

species reactivity

human

Gene Information

mouse ... Camk2a(12322)
rat ... Camk2a(25400)

Gene Information

rat ... Camk2a(25400)

Gene Information

human ... CAMK2D(817)

Gene Information

human ... CAMK2A(815), CAMK2B(816), CAMK2D(817)

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

biological source

mouse

biological source

mouse

biological source

rabbit

biological source

rabbit

clone

22B1, monoclonal

clone

6G9, monoclonal

clone

polyclonal

clone

polyclonal

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters