Skip to Content
Merck

Skip To

P2995

Anti-Phosphodiesterase 6α antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-PDE6α

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Please contact Customer Service for Availability
11 300,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-

11 300,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~90 kDa

species reactivity

bovine, sheep

technique(s)

western blot: 1-2 μg/mL using sheep retinal extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDE6A(5145)
mouse ... Pde6a(225600)

Immunogen

synthetic peptide corresponding to amino acid residues 1-16 from human PDE6α. This sequence is completely conserved in mouse and bovine and 94% conserved in canine.

Biochem/physiol Actions

Detects PDE6α from sheep retinal extract and bovine rod outer segment membranes as well as purified bovine PDE6.

Physical form

Solution in phosphate buffered saline containing 1 mg/mL bovine serum albumin and 0.05% sodium azide.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA074677MABN2250HPA016970
description

affinity isolated antibody, buffered aqueous solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

description

clone 7A11.1, from mouse

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

200

Quality Level

-

Quality Level

-

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

storage temp.

−20°C

form

buffered aqueous solution

form

buffered aqueous glycerol solution

form

-

form

buffered aqueous glycerol solution


Still not finding the right product?

— or —

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters