Skip to Content
Merck

P7771

Pyrimethamine

Synonym(s):

5-(4-Chlorophenyl)-6-ethyl-2,4-pyrimidinediamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H13ClN4
CAS Number:
Molecular Weight:
248.71
EC Number:
200-364-2
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
219864
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


SMILES string

CCC1=NC(N)=NC(N)=C1C2=CC=C(Cl)C=C2

InChI

1S/C12H13ClN4/c1-2-9-10(11(14)17-12(15)16-9)7-3-5-8(13)6-4-7/h3-6H,2H2,1H3,(H4,14,15,16,17)

InChI key

WKSAUQYGYAYLPV-UHFFFAOYSA-N

Gene Information



Still not finding the right product?

Explore all of our products under Pyrimethamine


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

WGK

WGK 3

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library