Skip to Content
Merck

Skip To

PLA0076

Rabbit anti-WAPL Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

FOE, KIAA0261, WAPAL, WAPL cohesin release factor, friend of EBNA2, friend of EBNA2 (Epstein-Barr virus nuclear protein 2), friend of EBNA2 protein, wings apart-like homolog (Drosophila)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Please contact Customer Service for Availability
9 270,00 CZK

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-

9 270,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: 1:10,000-1:25,000

accession no.

Q7Z5K2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... WAPL(23063)

Immunogen

The epitope recognized by PLA0076 maps to a region between residues 1125 and 1175 of human wings apart like/KIAA0261 using the numbering given in TrEMBL entry Q7Z5K2 (GeneID 23063).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Wings apart-like protein homolog (WAPL) is a cohesin binding protein that functions to destabilize cohesin-linked sister chromatids during prophase. Human WAPL has been shown to be highly expressed in cervical cancer and when over-expressed in mouse cells, induces tumorigenesis. WAPL was isolated in a screen for proteins that interact with Epstein-Barr nuclear protein 2 (EBNA2).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA037874HPA037875HPA066141
description

Powered by Bethyl Laboratories, Inc.

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

77-92-9

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters