Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μL | Please contact Customer Service for Availability | 12 700,00 CZK |
About This Item
12 700,00 CZK
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human, mouse
technique(s)
immunohistochemistry: 1:500- 1:2,000, immunoprecipitation (IP): 2-5 μg/mg, western blot: 1:1,000- 1:10,000
accession no.
NP_001008895.1
UniProt accession no.
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
rabbit ... Cul4a(8451)
Immunogen
Physical form
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description Powered by Bethyl Laboratories, Inc. | description clone 2A2.1, from mouse | description clone 23GB790, recombinant rabbit monoclonal, expressed in HEK293 cells | description Powered by Bethyl Laboratories, Inc. |
| biological source rabbit | biological source mouse | biological source rabbit (Monoclonal) | biological source rabbit |
| antibody form affinity purified immunoglobulin | antibody form purified immunoglobulin | antibody form affinity isolated antibody | antibody form affinity purified immunoglobulin |
| clone polyclonal | clone 2A2.1, monoclonal | clone 23GB790, recombinant monoclonal | clone polyclonal |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| species reactivity human, mouse | species reactivity human | species reactivity human | species reactivity human, mouse |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
77-92-9
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



