Skip to Content
Merck

Skip To

PLA0247

Rabbit anti-ARID4B Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

180 kDa Sin3-associated polypeptide, ARID domain-containing protein 4B, AT rich interactive domain 4B (RBP1- like), AT rich interactive domain 4B (RBP1-like), BCAA, BRCAA1, RBBP1L1, RBP1L1, Rb-binding protein homolog, SAP180, SIN3A-associated protein 180, breast cancer-associated antigen 1, breast cancer-associated antigen BRCAA1, breast carcinoma-associated antigen, histone deacetylase complex subunit SAP180, retinoblastoma binding protein 1-like 1, retinoblastoma-binding protein 1-like 1, sin3-associated polypeptide p180

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Please contact Customer Service for Availability
9 270,00 CZK

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
-

9 270,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunohistochemistry: 1:1,000- 1:5,000, immunoprecipitation (IP): 5-10 μg/mg, western blot: 1:2,000- 1:10,000

accession no.

NP_057458.4

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... ARID4B(51742)

Immunogen

The epitope recognized by PLA0247 maps to a region between residue 125 and 175 of human AT rich interactive domain 4B using the numbering given in entry NP_057458.4 (GeneID 51742).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

ARID4B (AT-rich interactive domain-containing protein 4B) is also known as RbBP1L1 (retinoblastoma binding protein 1-like 1) and is a retinoblastoma binding protein and member of the AT-rich interaction domain (ARID) family of proteins. In addition to the ARID DNA binding domain, ARID4B also contains a chromo domain and a TUDOR domain, indicating a function in chromatin organization and protein-protein interactions with methylated protein substrates. As a subunit of the SIN3A transcriptional co-repressor, ARID4B is implicated to be involved in chromatin remodeling and epigenetic modifications. ARID4B may play a role in the pathogenesis of breast cancer and other malignancies.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA027333HPA027205PLA0172
description

Powered by Bethyl Laboratories, Inc.

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

Powered by Bethyl Laboratories, Inc.

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity purified immunoglobulin

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Gene Information

rabbit ... ARID4B(51742)

Gene Information

human ... ARID4B(51742)

Gene Information

human ... ARID4B(51742)

Gene Information

rabbit ... PML(5371)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

77-92-9

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library