Skip to Content
Merck

Skip To

PLA0256

Rabbit anti-WDR5 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

BIG-3, BMP2-induced 3-kb gene protein, CFAP89, SWD3, Set1c WD40 repeat protein, cilia and flagella associated protein 89, homolog, homolog (S. cerevisiae)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Please contact Customer Service for Availability
12 700,00 CZK

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, PLA, WB
Citations:
1

12 700,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: 1:200- 1:1,000, proximity ligation assay: suitable, western blot: 1:2,000-1:10,000

accession no.

NP_060058.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... WDR5(11091)

Immunogen

The epitope recognized by PLA0256 maps to a region between residue 50 and 100 of human WD repeat domain 5 using the numbering given in entry NP_060058.1 (GeneID 11091).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

WD-repeat domain 5 (WDR5) is a member of the WD repeat family. The WD repeat is defined by four or more repeating units of a conserved core of approximately 40 amino acids ending with tryptophan-aspartic acid (WD). WD repeats may serve as sites of protein-protein interaction for adaptor proteins and facilitate multiprotein complex formation. WDR5 contains 7 WD repeats and is a core subunit of the human MLL and SET1 histone H3 lys4 methyltransferase complexes. WDR5 functions in facilitating the assembly of MLL and SET1 chromatin-modifiying complexes.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ZRB3079HPA047182HPA076612
description

Powered by Bethyl Laboratories, Inc.

description

recombinant, expressed in HEK 293 cells

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

clone

polyclonal

clone

3N4, recombinant monoclonal

clone

polyclonal

clone

polyclonal

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

species reactivity

human, mouse, rat

species reactivity

human, mouse, rat

species reactivity

human

species reactivity

human

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

-10 to -25°C


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters