Skip to Content
Merck

PLA0263

Rabbit anti-Ku70 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

5′-dRP lyase Ku70, 5′-deoxyribose-5-phosphate lyase Ku70, 70 kDa subunit, 70 kDa subunit of Ku antigen, 70kDa, ATP-dependent DNA helicase 2 subunit 1, ATP-dependent DNA helicase II, ATP-dependent DNA helicase II 70 kDa subunit, CTC box binding factor 75 kDa subunit, CTC box-binding factor 75 kDa subunit, CTC75, CTCBF, D22S671, D22S731, DNA repair protein XRCC6, G22P1, KU70, Ku autoantigen, Ku autoantigen p70 subunit, ML8, TLAA, Thyroid-lupus autoantigen, X-ray repair complementing defective repair in Chinese hamster cells 6, lupus Ku autoantigen protein p70, thyroid autoantigen 70kD (Ku antigen), thyroid autoantigen 70kDa (Ku antigen), thyroid-lupus autoantigen p70

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): 2-5 μg/mg, western blot: 1:2,000- 1:10,000

accession no.

NP_001460.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... Ku70(2547)

Immunogen

The epitope recognized by PLA0263 maps to a region between residue 500 and 550 of human Lupus Ku autoantigen protein p70 using the numbering given in entry NP_001460.1 (GeneID 2547).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

Ku70 is the 70 kDa subunit of the lupus Ku autoantigen. The lupus Ku autoantigen was identified in individuals with systemic lupus erythematosus (SLE) and related disorders. The Ku antigen is a heterodimer of Ku70 and Ku80. The Ku70/Ku80 complex functions as a single-stranded DNA-dependent ATP-dependent helicase. The complex may be involved in the repair of nonhomologous DNA ends such as that required for double-strand break repair, transposition, and V(D)J recombination. [taken from the Universal Protein Resource (UniProt) www.uniprot.org/uniprot/P12956].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library