Skip to Content
Merck

Skip To

PLA0290

Rabbit anti-Coilin Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

CLN80, coilin p80, p80, p80-coilin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Please contact Customer Service for Availability
12 700,00 CZK

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
-

12 700,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunohistochemistry: 1:500-1:2,000, immunoprecipitation (IP): 2- 10 μg/mg, western blot: 1:2,000- 1:10,000

accession no.

NP_004636.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... Coilin(8161)

Immunogen

The epitope recognized by PLA0290 maps to a region between residue 526 and 576 of human Coilin using the numbering given in entry NP_004636.1 (GeneID 8161).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Coilin is a component of the nuclear coiled bodies (CBS) which are involved in the function or assembly/disassembly of nucleoplasmic snRNPs. During mitosis, CBS disassemble, coinciding with a mitotic-specific phosphorylation of p80 coilin [taken from the Universal Protein Resource (UniProt) www.uniprot.org/uniprot/Q6PJP8].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C1862HPA068537SAB5702313
description

Powered by Bethyl Laboratories, Inc.

description

~1.5 mg/mL, clone pδ, purified from hybridoma cell culture

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

description

-

biological source

rabbit

biological source

mouse

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

mouse

antibody form

affinity purified immunoglobulin

antibody form

purified from hybridoma cell culture

antibody form

affinity isolated antibody

antibody form

-

clone

polyclonal

clone

pδ, monoclonal

clone

polyclonal

clone

7Z4K1, monoclonal

shipped in

wet ice

shipped in

dry ice

shipped in

wet ice

shipped in

-


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

77-92-9

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters