Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Please contact Customer Service for Availability | 14 500,00 CZK |
About This Item
sample type plasma
sample type cell culture supernatant(s)
14 500,00 CZK
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable, capture ELISA: suitable
input
sample type serum
sample type plasma
sample type cell culture supernatant(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 15 pg/mL
standard curve range: 24.69-18000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Ccl27(20301)
General description
The CCL27 (C-C motif chemokine ligand 27) gene codes for a T cell skin-homing chemokine and is expressed in epidermal keratinocytes. It is also referred to as cutaneous T cell-attracting chemokine (CTACK).
Immunogen
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Biochem/physiol Actions
Other Notes
Please type the word sample in the text box provided for lot number.
1 of 1
This Item | |||
|---|---|---|---|
| description for serum, plasma and cell culture supernatant | description for serum, plasma, cell culture supernatant and urine | description for serum, plasma and cell culture supernatant | description for serum, plasma and cell culture supernatant |
| species reactivity mouse | species reactivity human | species reactivity mouse | species reactivity mouse |
| Gene Information mouse ... Ccl27(20301) | Gene Information human ... CCL27(10850) | Gene Information mouse ... Ccl25(20300) | Gene Information mouse ... Ccl2(20296) |
| input sample type serum | input sample type serum | input sample type plasma | input sample type serum |
| assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| technique(s) ELISA: suitable, capture ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable |
| detection method colorimetric | detection method colorimetric | detection method colorimetric | detection method colorimetric |
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




