Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Please contact Customer Service for Availability | 19 600,00 CZK |
About This Item
sample type: human tissue lysate
19 600,00 CZK
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable, capture ELISA: suitable
input
sample type: human cell lysate
sample type: human tissue lysate
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... EGFR(1965)
General description
Immunogen
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
1 of 1
This Item | |||
|---|---|---|---|
| description for detection of human phospho-egfr (pTyr1086) and pan-egfr in cell and tissue lysates | description for detection of human phospho-egfr (pTyr992) and pan-egfr in cell and tissue lysates | description for detection of human phospho-egfr (pTyr1045) and pan-egfr in cell and tissue lysates | description for detection of human phospho-egfr (pTyr1045) in cell and tissue lysates |
| species reactivity human | species reactivity human | species reactivity human | species reactivity human |
| Gene Information human ... EGFR(1965) | Gene Information human ... EGFR(1965) | Gene Information human ... EGFR(1965) | Gene Information human ... EGFR(1965) |
| input sample type: human cell lysate | input sample type: human cell lysate | input sample type: human tissue lysate | input sample type: human cell lysate |
| detection method colorimetric | detection method - | detection method - | detection method - |
| technique(s) capture ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable | technique(s) capture ELISA: suitable |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - ED ENV 1 - Eye Irrit. 2 - Resp. Sens. 1
Storage Class
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




